4',5'-Dimethyl-2'-hydroxypropiophenon

Lieferant: CHEMPUR

FL12095.POR
CHMPFL12095.POREA 0 EUR
CHMPFL12095.POR
4',5'-Dimethyl-2'-hydroxypropiophenon
4',5'-Dimethyl-2'-hydroxypropiophenon
Formel: C₁₁H₁₄O₂
CAS-Nummer: 5384-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR