2'-Hydroxy-5'-methyl-3'-nitropropiophenon

Lieferant: CHEMPUR

FL12096.POR
CHMPFL12096.POREA 0 EUR
CHMPFL12096.POR
2'-Hydroxy-5'-methyl-3'-nitropropiophenon
2'-Hydroxy-5'-methyl-3'-nitropropiophenon
Formel: C₁₀H₁₁NO₄
CAS-Nummer: 70978-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR