5'-Ethyl-2'-hydroxy-3'-nitropropiophenon

Lieferant: CHEMPUR

FL12099.POR
CHMPFL12099.POREA 0 EUR
CHMPFL12099.POR
5'-Ethyl-2'-hydroxy-3'-nitropropiophenon
5'-Ethyl-2'-hydroxy-3'-nitropropiophenon
Formel: C₁₁H₁₃NO₄
CAS-Nummer: 70978-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR