2'-Hydroxy-5'-isopropyl-3'-nitropropiophenon

Lieferant: CHEMPUR

FL12100.POR
CHMPFL12100.POREA 0 EUR
CHMPFL12100.POR
2'-Hydroxy-5'-isopropyl-3'-nitropropiophenon
2'-Hydroxy-5'-isopropyl-3'-nitropropiophenon
Formel: C₁₂H₁₅NO₄
CAS-Nummer: 288401-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR