6-Methylflavon

Lieferant: CHEMPUR

Synonyme: 6-Methyl-2-phenyl-4H-chromen-4-on, 6-Methyl-2-phenylchromen-4-on, 6-Methyl-2-phenyl-4H-1-benzopyran-4-on, 6-Methyl-2-phenyl-4H-benzo[b]pyran-4-on, 6-Methyl-2-phenyl-4-benzopyron, 6-Methyl-2-phenylchromon

FL12102.POR
CHMPFL12102.POREA 0 EUR
CHMPFL12102.POR
6-Methylflavon
6-Methylflavon
Formel: C₁₆H₁₂O₂
Molecular Weight: 236,27 g/mol
MDL: MFCD00017461
CAS-Nummer: 29976-75-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR