6-Chlor-7-methylflavon

Lieferant: CHEMPUR

Synonyme: 6-Chlor-7-methyl-2-phenylchromen-4-on, 6-Chlor-7-methyl-2-phenyl-4H-chromen-4-on, 6-Chlor-7-methyl-2-phenyl-4H-1-benzopyran-4-on, 6-Chlor-7-methyl-2-phenyl-4H-benzo[b]pyran-4-on, 6-Chlor-7-methyl-2-phenylchromon, 6-Chlor-7-methyl-2-phenyl-4-benzopyron

FL12105.POR
CHMPFL12105.POREA 0 EUR
CHMPFL12105.POR
6-Chlor-7-methylflavon
6-Chlor-7-methylflavon
Formel: C₁₆H₁₁ClO₂
Molecular Weight: 270,71 g/mol
CAS-Nummer: 147919-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR