7-Hydroxyflavon

Lieferant: CHEMPUR

Synonyme: 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-on, 7-Hydroxy-2-phenyl-4-benzopyron, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-on, 7-Hydroxy-2-phenylchromen-4-on, 7-Hydroxy-2-phenyl-4H-chromen-4-on

FL12111.POR
CHMPFL12111.POREA 0 EUR
CHMPFL12111.POR
7-Hydroxyflavon
7-Hydroxyflavon
Formel: C₁₅H₁₀O₃
Molecular Weight: 238,24 g/mol
Siedepunkt: 395 °C (1013 hPa)
Schmelzpunkt: 244…244,5 °C
MDL: MFCD00006835
CAS-Nummer: 6665-86-7
EINECS: 229-705-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR