6-Chlor-4-chromanon

Lieferant: CHEMPUR

CFF588.1 FL12139.POR
CHMPCFF588.1EA 532 EUR
CHMPCFF588.1 CHMPFL12139.POR
6-Chlor-4-chromanon
6-Chlor-4-chromanon
Formel: C₉H₇ClO₂
Molecular Weight: 182,61 g/mol
MDL: MFCD00024070
CAS-Nummer: 37674-72-9
EINECS: 253-611-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR