6-Brom-1,4-benzodioxan

Lieferant: CHEMPUR

Synonyme: 6-Bromo-2,3-dihydro-1,4-benzodioxine, 6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine

FL12237.POR
CHMPFL12237.POREA 0 EUR
CHMPFL12237.POR
6-Brom-1,4-benzodioxan
6-Brom-1,4-benzodioxan
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD00040750
CAS-Nummer: 52287-51-1
EINECS: 257-817-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR