1,2,3,4-Tetrahydronaphthalin-2-on (2-Tetralon, β-Tetralon)

Lieferant: CHEMPUR

Synonyme: Beta-tetralone

FL12252.POR
CHMPFL12252.POREA 0 EUR
CHMPFL12252.POR
1,2,3,4-Tetrahydronaphthalin-2-on (2-Tetralon, β-Tetralon)
1,2,3,4-Tetrahydronaphthalin-2-on (2-Tetralon, β-Tetralon)
Formel: C₁₀H₁₀O
Molecular Weight: 146,19 g/mol
MDL: MFCD00001727
CAS-Nummer: 530-93-8
EINECS: 208-498-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR