3-(4-Methylphenoxy)propionsäure

Lieferant: CHEMPUR

Synonyme: 3-(p-Tolyloxy)propanoic acid

FL12265.POR
CHMPFL12265.POREA 0 EUR
CHMPFL12265.POR
3-(4-Methylphenoxy)propionsäure
3-(4-Methylphenoxy)propionsäure
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
MDL: MFCD00220218
CAS-Nummer: 25173-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR