3-(4-Chlorphenoxy)propionsäure

Lieferant: CHEMPUR

Synonyme: 3-(4-Chlorophenoxy)propanoic acid

CFF972.1 FL12266.POR
CHMPCFF972.1EA 522 EUR
CHMPCFF972.1 CHMPFL12266.POR
3-(4-Chlorphenoxy)propionsäure
3-(4-Chlorphenoxy)propionsäure
Formel: C₉H₉ClO₃
Molecular Weight: 200,6190 g/mol
MDL: MFCD00043504
CAS-Nummer: 3284-79-5
EINECS: 221-932-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR