2-Iod-4-(trifluormethoxy)phenol

Lieferant: CHEMPUR

Synonyme: 1-Hydroxy-2-iod-4-(trifluormethoxy)benzol

FL13046.POR
CHMPFL13046.POREA 0 EUR
CHMPFL13046.POR
2-Iod-4-(trifluormethoxy)phenol
2-Iod-4-(trifluormethoxy)phenol
Formel: C₇H₄F₃IO₂
CAS-Nummer: 405517-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR