2-Fluor-5-iodbenzoylchlorid

Lieferant: CHEMPUR

FL13055.POR
CHMPFL13055.POREA 0 EUR
CHMPFL13055.POR
2-Fluor-5-iodbenzoylchlorid
2-Fluor-5-iodbenzoylchlorid
Formel: C₇H₃ClFIO
MDL: MFCD03094145
CAS-Nummer: 186584-73-6
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR