3-Ethoxybenzoylchlorid

Lieferant: CHEMPUR

FL13056.POR
CHMPFL13056.POREA 0 EUR
CHMPFL13056.POR
3-Ethoxybenzoylchlorid
3-Ethoxybenzoylchlorid
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
MDL: MFCD00000677
CAS-Nummer: 61956-65-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR