(3-Fluor-4-hydroxyphenyl)essigsäure

Lieferant: CHEMPUR

FL13059.POR 013059-1G 013059-25G 013059-5G
CHMPFL13059.POREA 0 EUR
CHMPFL13059.POR FLUO013059-1G FLUO013059-25G FLUO013059-5G
(3-Fluor-4-hydroxyphenyl)essigsäure
(3-Fluor-4-hydroxyphenyl)essigsäure
Formel: C₈H₇FO₃
MDL: MFCD00004348
CAS-Nummer: 458-09-3
EINECS: 207-275-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR