(2-Chlor-4-fluorphenyl)essigsäure

Lieferant: CHEMPUR
Achtung

Synonyme: 2-(2-Chloro-4-fluorophenyl)acetic acid

FL13061.POR
CHMPFL13061.POREA 0 EUR
CHMPFL13061.POR
(2-Chlor-4-fluorphenyl)essigsäure
(2-Chlor-4-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Molecular Weight: 188,59 g/mol
Siedepunkt: 289 °C (1013 hPa)
Schmelzpunkt: 101…104 °C
MDL: MFCD00236028
CAS-Nummer: 177985-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR