2-(Trifluormethoxy)phenylacetonitril

Lieferant: CHEMPUR

Synonyme: 2-(Trifluormethoxy)benzylcyanid

FL13062.5
CHMPFL13062.5EA 0 EUR
CHMPFL13062.5
2-(Trifluormethoxy)phenylacetonitril
2-(Trifluormethoxy)phenylacetonitril
Formel: C₉H₆F₃NO
Molecular Weight: 201,15 g/mol
MDL: MFCD00236326
CAS-Nummer: 137218-25-8
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR