6-Nitro-2,3,5-trifluoranilin

Lieferant: CHEMPUR

Synonyme: 2-Nitro-3,5,6-trifluoranilin

FL13089.POR
CHMPFL13089.POREA 0 EUR
CHMPFL13089.POR
6-Nitro-2,3,5-trifluoranilin
6-Nitro-2,3,5-trifluoranilin
Formel: C₆H₃F₃N₂O₂
Molecular Weight: 192,1 g/mol
MDL: MFCD00971838
CAS-Nummer: 5415-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR