2-Brom-5-chlorbenzotrifluorid

Lieferant: CHEMPUR

FL13188.POR 013188-100G 013188-25G
CHMPFL13188.POREA 0 EUR
CHMPFL13188.POR FLUO013188-100G FLUO013188-25G
2-Brom-5-chlorbenzotrifluorid
2-Brom-5-chlorbenzotrifluorid
Formel: C₇H₃BrClF₃
MDL: MFCD00010308
CAS-Nummer: 344-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR