2-Brom-6-chlor-4-fluoranilin
Lieferant: CHEMPUR
FL13190.POR
013190-5G
013190-100G
CHMPFL13190.POREA
0
EUR
CHMPFL13190.POR
FLUO013190-5G
FLUO013190-100G
2-Brom-6-chlor-4-fluoranilin
2-Brom-6-chlor-4-fluoranilin
Formel:
C₆H₄BrClFN Siedepunkt: 260 °C (1013 hPa) Schmelzpunkt: 55…56 °C |
MDL:
MFCD00051707 CAS-Nummer: 201849-14-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...