2-Brom-4,5-difluoranisol

Lieferant: CHEMPUR
Achtung

Synonyme: 1-Bromo-4,5-difluoro-2-methoxybenzene

FL13196.POR
CHMPFL13196.POREA 0 EUR
CHMPFL13196.POR
2-Brom-4,5-difluoranisol
2-Brom-4,5-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223,02 g/mol
MDL: MFCD00070753
CAS-Nummer: 202865-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR