2-Brom-4,6-difluorphenol

Lieferant: CHEMPUR
Achtung

CFF836. FL13197.POR 013197-25G 013197-5G
CHMPCFF836.EA 0 EUR
CHMPCFF836. CHMPFL13197.POR FLUO013197-25G FLUO013197-5G
2-Brom-4,6-difluorphenol
2-Brom-4,6-difluorphenol
Formel: C₆H₃BrF₂O
MDL: MFCD00070752
CAS-Nummer: 98130-56-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR