2'-Brom-4'-fluoracetanilid

Lieferant: CHEMPUR

Synonyme: N-(2-Brom-4-fluorphenyl)acetamid

FL13199.POR
CHMPFL13199.POREA 0 EUR
CHMPFL13199.POR
2'-Brom-4'-fluoracetanilid
2'-Brom-4'-fluoracetanilid
Formel: H₃CC(O)NH(C₆H₃BrF)
Molecular Weight: 232,05 g/mol
Siedepunkt: 344 °C (1013 hPa)
Schmelzpunkt: 119…120 °C
MDL: MFCD00061118
CAS-Nummer: 1009-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR