4'-Brom-3'-fluoracetanilid

Lieferant: CHEMPUR

Synonyme: N-(4-Brom-3-fluorphenyl)acetamid

FL13200.POR
CHMPFL13200.POREA 0 EUR
CHMPFL13200.POR
4'-Brom-3'-fluoracetanilid
4'-Brom-3'-fluoracetanilid
Formel: H₃CC(O)NH(C₆H₃BrF)
Molecular Weight: 232,05 g/mol
MDL: MFCD00672937
CAS-Nummer: 351-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR