4'-Brom-2'-fluoracetophenon

Lieferant: CHEMPUR

Synonyme: 1-(4-Bromo-2-fluorophenyl)ethanone

FL13201.POR
CHMPFL13201.POREA 0 EUR
CHMPFL13201.POR
4'-Brom-2'-fluoracetophenon
4'-Brom-2'-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217,04 g/mol
MDL: MFCD03411551
CAS-Nummer: 625446-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR