2-Brom-4-fluorbenzylalkohol

Lieferant: CHEMPUR

Synonyme: (2-Brom-4-fluorphenyl)methanol

FL13215.POR
CHMPFL13215.POREA 0 EUR
CHMPFL13215.POR
2-Brom-4-fluorbenzylalkohol
2-Brom-4-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD00672925
CAS-Nummer: 229027-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR