5-Brom-2-fluorbenzylalkohol

Lieferant: CHEMPUR

Synonyme: (5-Bromo-2-fluorophenyl)methanol

FL13220.POR
CHMPFL13220.POREA 0 EUR
CHMPFL13220.POR
5-Brom-2-fluorbenzylalkohol
5-Brom-2-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD00143425
CAS-Nummer: 99725-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR