2-Brom-5-fluorzimtsäure

Lieferant: CHEMPUR
Achtung

Synonyme: 3-(2-Brom-5-fluorphenyl)propensäure, 2-Brom-5-fluorcinnamsäure

FL13230.POR
CHMPFL13230.POREA 0 EUR
CHMPFL13230.POR
2-Brom-5-fluorzimtsäure
2-Brom-5-fluorzimtsäure
Formel: C₉H₆BrFO₂
MDL: MFCD00142878
CAS-Nummer: 202865-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR