3-Brom-5-fluorzimtsäure

Lieferant: CHEMPUR

FL13231.POR
CHMPFL13231.POREA 0 EUR
CHMPFL13231.POR
3-Brom-5-fluorzimtsäure
3-Brom-5-fluorzimtsäure
Formel: C₉H₆BrFO₂
Molecular Weight: 245,05 g/mol
CAS-Nummer: 887268-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR