4-Brom-2-fluor-5-iodtoluol

Lieferant: CHEMPUR

Synonyme: 1-Brom-3-fluor-6-iod-4-methylbenzol

FL13235.POR
CHMPFL13235.POREA 0 EUR
CHMPFL13235.POR
4-Brom-2-fluor-5-iodtoluol
4-Brom-2-fluor-5-iodtoluol
Formel: C₇H₅BrFI
Molecular Weight: 314,92 g/mol
MDL: MFCD06200719
CAS-Nummer: 870704-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR