5-Brom-2-fluor-4-iodtoluol

Lieferant: CHEMPUR

Synonyme: 1-Brom-4-fluor-2-iod-5-methylbenzol

FL13236.POR
CHMPFL13236.POREA 0 EUR
CHMPFL13236.POR
5-Brom-2-fluor-4-iodtoluol
5-Brom-2-fluor-4-iodtoluol
Formel: C₇H₅BrFI
Molecular Weight: 314,92 g/mol
MDL: MFCD00143260
CAS-Nummer: 202865-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR