3'-Brom-4'-fluorpropiophenon

Lieferant: CHEMPUR

FL13245.POR
CHMPFL13245.POREA 0 EUR
CHMPFL13245.POR
3'-Brom-4'-fluorpropiophenon
3'-Brom-4'-fluorpropiophenon
Formel: C₉H₈BrFO
MDL: MFCD00143012
CAS-Nummer: 202865-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR