4'-Brom-2'-(trifluormethyl)acetanilid

Lieferant: CHEMPUR

FL13248.POR
CHMPFL13248.POREA 0 EUR
CHMPFL13248.POR
4'-Brom-2'-(trifluormethyl)acetanilid
4'-Brom-2'-(trifluormethyl)acetanilid
Formel: C₉H₇BrF₃NO
MDL: MFCD00672936
CAS-Nummer: 29124-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR