4'-Butoxy-2'-fluoracetophenon

Lieferant: CHEMPUR

FL13249.POR
CHMPFL13249.POREA 0 EUR
CHMPFL13249.POR
4'-Butoxy-2'-fluoracetophenon
4'-Butoxy-2'-fluoracetophenon
Formel: C₁₂H₁₅FO₂
MDL: MFCD00143201
CAS-Nummer: 203066-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR