2'-Chlor-4',5'-difluoracetophenon

Lieferant: CHEMPUR

Synonyme: 1-(2-Chloro-4,5-difluorophenyl)ethanone, 1-chloro-3,4-difluor-6-ethanoylbenzene

FL13250.POR
CHMPFL13250.POREA 0 EUR
CHMPFL13250.POR
2'-Chlor-4',5'-difluoracetophenon
2'-Chlor-4',5'-difluoracetophenon
Formel: C₈H₅ClF₂O
Molecular Weight: 190,58 g/mol
MDL: MFCD00671761
CAS-Nummer: 121872-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR