3'-Chlor-4'-fluoracetanilid

Lieferant: CHEMPUR

Synonyme: N-(3-Chlor-4-fluorphenyl)acetamid

FL13254.POR
CHMPFL13254.POREA 0 EUR
CHMPFL13254.POR
3'-Chlor-4'-fluoracetanilid
3'-Chlor-4'-fluoracetanilid
Formel: C₈H₇ClFNO
Molecular Weight: 187,6 g/mol
MDL: MFCD00018095
CAS-Nummer: 877-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR