3-Chlor-2-fluoranisol

Lieferant: CHEMPUR

Synonyme: 1-Chloro-2-fluoro-3-methoxybenzene

FL13257.POR
CHMPFL13257.POREA 0 EUR
CHMPFL13257.POR
3-Chlor-2-fluoranisol
3-Chlor-2-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160.58 g/mol
MDL: MFCD01631570
CAS-Nummer: 261762-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR