4-Chlor-2-fluoranisol

Lieferant: CHEMPUR
Achtung

Synonyme: 4-Chloro-2-fluoro-1-methoxybenzene

FL13259.POR
CHMPFL13259.POREA 0 EUR
CHMPFL13259.POR
4-Chlor-2-fluoranisol
4-Chlor-2-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD00672964
CAS-Nummer: 452-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR