5-Chlor-2-fluoranisol

Lieferant: CHEMPUR

Synonyme: 4-Chloro-1-fluoro-2-methoxybenzene

FL13260.POR
CHMPFL13260.POREA 0 EUR
CHMPFL13260.POR
5-Chlor-2-fluoranisol
5-Chlor-2-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160.58
CAS-Nummer: 1092349-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR