4-Chlor-3-fluorbenzonitril

Lieferant: CHEMPUR

CFF942. FL13264.POR
CHMPCFF942.EA 0 EUR
CHMPCFF942. CHMPFL13264.POR
4-Chlor-3-fluorbenzonitril
4-Chlor-3-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
MDL: MFCD00143291
CAS-Nummer: 110888-15-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR