4-Chlor-3-fluorbenzylalkohol

Lieferant: CHEMPUR

FL13265.POR
CHMPFL13265.POREA 0 EUR
CHMPFL13265.POR
4-Chlor-3-fluorbenzylalkohol
4-Chlor-3-fluorbenzylalkohol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD00143292
CAS-Nummer: 202925-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR