2-Chlor-4-fluorzimtsäure

Lieferant: CHEMPUR
Achtung

Synonyme: 3-(2-Chlor-4-fluorphenyl)propensäure, 2-Chlor-4-fluorcinnamsäure

FL13270.POR
CHMPFL13270.POREA 0 EUR
CHMPFL13270.POR
2-Chlor-4-fluorzimtsäure
2-Chlor-4-fluorzimtsäure
Formel: C₉H₆ClFO₂
Molecular Weight: 200,6 g/mol
Schmelzpunkt: 245…247 °C
MDL: MFCD00236027
CAS-Nummer: 133220-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR