4-Chlor-2-fluorzimtsäure

Lieferant: CHEMPUR
Achtung

Synonyme: 3-(4-Chlor-2-fluorphenyl)propensäure, 4-Chlor-2-fluorcinnamsäure

FL13271.POR
CHMPFL13271.POREA 0 EUR
CHMPFL13271.POR
4-Chlor-2-fluorzimtsäure
4-Chlor-2-fluorzimtsäure
Formel: C₉H₆ClFO₂
Molecular Weight: 200,6 g/mol
MDL: MFCD00143287
CAS-Nummer: 202982-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR