5-Chlor-2-fluorphenol

Lieferant: CHEMPUR

FL13285.POR
CHMPFL13285.POREA 0 EUR
CHMPFL13285.POR
5-Chlor-2-fluorphenol
5-Chlor-2-fluorphenol
Formel: C₆H₄ClFO
Molecular Weight: 146,55 g/mol
MDL: MFCD04116357
CAS-Nummer: 186589-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR