4-Chlor-3-fluortoluol

Lieferant: CHEMPUR

FL13287.POR 013287-5G
CHMPFL13287.POREA 0 EUR
CHMPFL13287.POR FLUO013287-5G
4-Chlor-3-fluortoluol
4-Chlor-3-fluortoluol
Formel: C₇H₆ClF
Molecular Weight: 144.58 g/mol
MDL: MFCD00143295
CAS-Nummer: 5527-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR