2,4-Dibrom-6-fluoranilin

Lieferant: CHEMPUR

FL13291.POR
CHMPFL13291.POREA 0 EUR
CHMPFL13291.POR
2,4-Dibrom-6-fluoranilin
2,4-Dibrom-6-fluoranilin
Formel: C₆H₄Br₂FN
Molecular Weight: 268,91 g/mol
Siedepunkt: 277 °C (1013 hPa)
Schmelzpunkt: 62…64 °C
MDL: MFCD00042230
CAS-Nummer: 141474-37-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR