2,4-Dibrom-6-fluoranisol

Lieferant: CHEMPUR

Synonyme: 1,3-Dibrom-5-fluor-4-methoxybenzol

FL13292.POR
CHMPFL13292.POREA 0 EUR
CHMPFL13292.POR
2,4-Dibrom-6-fluoranisol
2,4-Dibrom-6-fluoranisol
Formel: C₇H₅Br₂FO
Molecular Weight: 283,92 g/mol
MDL: MFCD00070788
CAS-Nummer: 202982-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR