2,6-Dibrom-4-fluoranisol

Lieferant: CHEMPUR

FL13293.POR
CHMPFL13293.POREA 0 EUR
CHMPFL13293.POR
2,6-Dibrom-4-fluoranisol
2,6-Dibrom-4-fluoranisol
Formel: C₇H₅Br₂FO
MDL: MFCD00042231
CAS-Nummer: 443-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR