2,4-Dibrom-6-fluorphenol

Lieferant: CHEMPUR
Achtung

Synonyme: 1,3-Dibromo-5-fluoro-4-hydroxybenzene

FL13294.POR
CHMPFL13294.POREA 0 EUR
CHMPFL13294.POR
2,4-Dibrom-6-fluorphenol
2,4-Dibrom-6-fluorphenol
Formel: C₆H₃Br₂FO
Molecular Weight: 269,9 g/mol
MDL: MFCD00042233
CAS-Nummer: 576-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR